accessibility menu, dialog, popup

UserName

CAS RN 24038-68-4

IUPAC Name: 5-(2-{6-hydroxy-[1,1'-biphenyl]-3-yl}propan-2-yl)-[1,1'-biphenyl]-2-ol
Synonyms: 5-(2-{6-hydroxy-[1,1'-biphenyl]-3-yl}propan-2-yl)-[1,1'-biphenyl]-2-ol
Molecular Weight (g/mol): 380.49
Molecular Formula: C27H24O2
InChi Key: BKTRENAPTCBBFA-UHFFFAOYSA-N
SMILES: CC(C)(C1=CC(=C(O)C=C1)C1=CC=CC=C1)C1=CC(=C(O)C=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results