CAS RN 2421153-77-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ (+)-JQ1 bump, acid functionalized
Functionalized BET bromodomain ligand; precursor for Tag Degraders