accessibility menu, dialog, popup

UserName

CH3H3CHONOO

CAS RN 2423-71-4

IUPAC Name: 2,6-dimethyl-4-nitrophenol
Synonyms: 2,6-dimethyl-4-nitrophenol
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: FNORUNUDZNWQFF-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)