accessibility menu, dialog, popup

UserName

OO

CAS RN 2426-07-5

IUPAC Name: 2-[4-(oxiran-2-yl)butyl]oxirane
Synonyms: 1,2,7,8-diepoxyoctane 1,7-octadiene diepoxide ccris 963 2,2'-butane-1,4-diyldioxirane
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: LFKLPJRVSHJZPL-UHFFFAOYNA-N
SMILES: C(CCC1CO1)CC1CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results