CAS RN 24353-58-0
CAS RN 24353-58-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Delachlor, TRC
CAS: 24353-58-0 Molecular Formula: C15 H22 Cl N O2 Molecular Weight (g/mol): 283.79 Synonym: 2',6'-Acetoxylidide, 2-chloro-N-(isobutoxymethyl)- (8CI),2-Chloro-N-(2,6-dimethylphenyl)-N-[(2-methylpropoxy)methyl]acetamide,2-Chloro-2',6'-dimethyl-N-(isobutoxymethyl)acetanilide,2-Chloro-N-(isobutoxymethyl)-2',6'-acetoxylidide,2-Chloro-N-(isobutoxymethyl)-2',6'-dimethylacetanilide,CP 52223,Delachlor,NSC 221681 IUPAC Name: 2-chloro-N-(2,6-dimethylphenyl)-N-(2-methylpropoxymethyl)acetamide SMILES: CC(C)COCN(C(=O)CCl)c1c(C)cccc1C