missing translation for 'onlineSavingsMsg'
Learn More

Delachlor, TRC

Product Code. 30366493
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366493 25 mg 25mg
30349807 100 mg 100mg
30361319 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366493 Supplier Toronto Research Chemicals Supplier No. TRCD23067625MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24353-58-0
Molecular Formula C15 H22 Cl N O2
Molecular Weight (g/mol) 283.79
Synonym 2',6'-Acetoxylidide, 2-chloro-N-(isobutoxymethyl)- (8CI), 2-Chloro-N-(2,6-dimethylphenyl)-N-[(2-methylpropoxy)methyl]acetamide, 2-Chloro-2',6'-dimethyl-N-(isobutoxymethyl)acetanilide, 2-Chloro-N-(isobutoxymethyl)-2',6'-acetoxylidide, 2-Chloro-N-(isobutoxymethyl)-2',6'-dimethylacetanilide, CP 52223, Delachlor, NSC 221681
IUPAC Name 2-chloro-N-(2,6-dimethylphenyl)-N-(2-methylpropoxymethyl)acetamide
SMILES CC(C)COCN(C(=O)CCl)c1c(C)cccc1C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H22ClNO2/c1-11(2)9-19-10-17(14(18)8-16)15-12(3)6-5-7-13(15)4/h5-7,11H,8-10H2,1-4H3
Formula Weight 283.1339
Chemical Name or Material Delachlor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.