accessibility menu, dialog, popup

UserName

CAS RN 2437-49-2

OHBrBrHOBr

CAS RN 2437-49-2

IUPAC Name: 2,4,6-tribromobenzene-1,3-diol
Synonyms: 2,4,6-tribromoresorcinol
Molecular Weight (g/mol): 346.8
Molecular Formula: C6H3Br3O2
InChi Key: YADZSMVDNYXOOB-UHFFFAOYSA-N
SMILES: OC1=C(Br)C(O)=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results