CAS RN 244105-55-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2'-O-(2-Methoxyethyl)-5-methylcytidine (~90%), TRC
CAS: 244105-55-3 Molecular Formula: C13H21N3O6 Molecular Weight (g/mol): 315.32 Synonym: 4-Amino-1-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one IUPAC Name: 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)oxolan-2-yl]-5-methylpyrimidin-2-one SMILES: COCCO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C=C(C)C(=NC2=O)N