missing translation for 'onlineSavingsMsg'
Learn More

2'-O-(2-Methoxyethyl)-5-methylcytidine (~90%), TRC

Product Code. 30476660
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476660 100 mg 100mg
30476658 250 mg 250mg
30466206 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476660 Supplier Toronto Research Chemicals Supplier No. TRCM304510100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244105-55-3
Molecular Formula C13H21N3O6
Molecular Weight (g/mol) 315.32
Synonym 4-Amino-1-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one
IUPAC Name 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)oxolan-2-yl]-5-methylpyrimidin-2-one
SMILES COCCO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C=C(C)C(=NC2=O)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H21N3O6/c1-7-5-16(13(19)15-11(7)14)12-10(21-4-3-20-2)9(18)8(6-17)22-12/h5,8-10,12,17-18H,3-4,6H2,1-2H3,(H2,14,15,19)/t8-,9-,10-,12-/m1/s1
Formula Weight 315.143
Chemical Name or Material 2'-O-(2-Methoxyethyl)-5-methylcytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.