accessibility menu, dialog, popup

UserName

CH3SSO

CAS RN 24445-35-0

IUPAC Name: 5-(methylsulfanyl)thiophene-2-carbaldehyde
Synonyms: 5-(methylsulfanyl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 158.23
Molecular Formula: C6H6OS2
InChi Key: RKXYTFTZODXDEF-UHFFFAOYSA-N
SMILES: CSC1=CC=C(S1)C=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results