missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylthio)thiophene-2-carbaldehyde, 90%, Thermo Scientific™

Product Code. 10651693
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10651693 10 g Amber Glass Bottle 10g
10436292 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10651693 Supplier Thermo Scientific Maybridge Supplier No. SEW03493EA
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 24445-35-0
Molecular Formula C6H6OS2
Molecular Weight (g/mol) 158.233
MDL Number MFCD00173744
InChI Key RKXYTFTZODXDEF-UHFFFAOYSA-N
Synonym 5-methylthio thiophene-2-carbaldehyde, 5-methylthio thiophene-2-carboxaldehyde, 5-methylsulfanyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-methylthio, 2-formyl-5-methylthiothiphene, 2-formyl-5-methylthio thiophene, 5-methylmercapto-2-thiophenaldehyde, 5-methylthio-2-thiophenecarbaldehyde, 5-methylthiothiophene-2-carbaldehyde, 5-methylmercaptothiophene-2-carbaldehyde
PubChem CID 2736997
IUPAC Name 5-methylsulfanylthiophene-2-carbaldehyde
SMILES CSC1=CC=C(S1)C=O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 158.24
Percent Purity 90%
Chemical Name or Material 5-(Methylthio)thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.