accessibility menu, dialog, popup

UserName

CAS RN 251320-85-1

IUPAC Name: dicyclohexyl[2'-(propan-2-yl)-[1,1'-biphenyl]-2-yl]phosphane
Synonyms: dicyclohexyl({2'-isopropyl-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol): 392.57
Molecular Formula: C27H37P
InChi Key: PQWWLQRSCJJOIS-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results