CAS RN 25173-76-6
CAS RN 25173-76-6
IUPAC Name:
3-(3,4-dimethylphenyl)propanoic acid
Synonyms:
3-(3,4-dimethylphenyl)propanoic acid
Molecular Weight (g/mol):
178.23
Molecular Formula:
C11H14O2
InChi Key:
FHEOAGCWJWUKJJ-UHFFFAOYSA-N
SMILES:
CC1=C(C)C=C(CCC(O)=O)C=C1
1
–
1
of
1
results
1
3-(3,4-Dimethylphenyl)propionic acid, 96%, Thermo Scientific™
CAS: 25173-76-6 Molecular Formula: C11H14O2 Molecular Weight (g/mol): 178.23 MDL Number: MFCD03425686 InChI Key: FHEOAGCWJWUKJJ-UHFFFAOYSA-N PubChem CID: 3870218 IUPAC Name: 3-(3,4-dimethylphenyl)propanoic acid SMILES: CC1=C(C)C=C(CCC(O)=O)C=C1