accessibility menu, dialog, popup

UserName

CAS RN 251945-92-3

NHNNHNHO

CAS RN 251945-92-3

IUPAC Name: 4-({2-phenyl-7H-pyrrolo[2,3-d]pyrimidin-4-yl}amino)cyclohexan-1-ol
Synonyms: 4-({2-phenyl-7H-pyrrolo[2,3-d]pyrimidin-4-yl}amino)cyclohexan-1-ol
Molecular Weight (g/mol): 308.39
Molecular Formula: C18H20N4O
InChi Key: RBZNJGHIKXAKQE-UHFFFAOYSA-N
SMILES: OC1CCC(CC1)NC1=C2C=CNC2=NC(=N1)C1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results