missing translation for 'onlineSavingsMsg'
Learn More

Derenofylline, MedChemExpress

Product Code. 16496208
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496208 10 mg 10mg
16438807 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496208 Supplier Medchem Express Supplier No. HY14858
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Derenofylline (SLV 320) is a potent, selective and orally active adenosine A1 receptor antagonist, with Ki values of 1 nM, 200 nM and 398 nM for human A1, A3 and A2A receptors respectively. Derenofylline suppresses cardiac fibrosis and attenuates albuminuria without affecting blood pressure in rats.

Derenofylline (SLV 320) is a potent, selective and orally active adenosine A1 receptor antagonist, with Ki values of 1 nM, 200 nM and 398 nM for human A1, A3 and A2A receptors respectively. Derenofylline suppresses cardiac fibrosis and attenuates albuminuria without affecting blood pressure in rats.

Specifications

Chemical Name or Material Derenofylline
CAS 251945-92-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H20N4O
Quantity 10 mg
Synonym SLV 320
Solubility Information DMSO : 250 mg/mL (810.69 mM; Need ultrasonic)
SMILES O[C@H]1CC[C@H](NC2=C3C(NC=C3)=NC(C4=CC=CC=C4)=N2)CC1
Molecular Weight (g/mol) 308.38
Formula Weight 308.38
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.