accessibility menu, dialog, popup

UserName

CAS RN 254993-59-4

IUPAC Name: [2-chloro-4-(trifluoromethyl)phenyl]boronic acid
Synonyms: 2-chloro-4-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 224.37
Molecular Formula: C7H5BClF3O2
InChi Key: JKSUCAFAUNSPLO-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results