accessibility menu, dialog, popup

UserName

CAS RN 2553-04-0

CH3OO

CAS RN 2553-04-0

IUPAC Name: (2-methoxyphenyl)(phenyl)methanone
Synonyms: (2-methoxyphenyl)(phenyl)methanone
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: CSUUDNFYSFENAE-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results