accessibility menu, dialog, popup

UserName

CAS RN 2567-83-1

IUPAC Name: tetraethylazanium perchlorate
Synonyms: tetraethylammonium perchlorate
Molecular Weight (g/mol): 229.7
Molecular Formula: C8H20ClNO4
InChi Key: WGHUNMFFLAMBJD-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results