missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium perchlorate, 0.2M aqueous solution

Product Code. 10483755 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Plastic bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10483755 100 mL Plastic bottle 100mL
10350634 500 mL Plastic bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10483755 Supplier Thermo Scientific Acros Supplier No. 379801000
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2567-83-1 | C8H20ClNO4 | 229.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraethylammonium perchlorate
CAS 2567-83-1, 7732-18-5
Density 1.0016g/mL
Packaging Plastic bottle
Molecular Formula C8H20ClNO4
MDL Number MFCD00041977
Quantity 100 mL
Specific Gravity 1.0016
Synonym tetraethylammonium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate, tetraethylazanium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate 1:1, ammonium, tetraethyl-, perchlorate, tetraethylammoniumperchlorate, acmc-1ccq7, ksc203k4p, tetraethyl ammonium perchlorate, tetraethylammoniumperchlorate, dry
InChI Key WGHUNMFFLAMBJD-UHFFFAOYSA-M
SMILES [O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC
IUPAC Name tetraethylazanium;perchlorate
Molecular Weight (g/mol) 229.70
PubChem CID 2734932
Formula Weight 229.7
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.