accessibility menu, dialog, popup

UserName

H3CO

CAS RN 25870-62-6

IUPAC Name: 1-phenylhexan-2-one
Synonyms: 1-phenylhexan-2-one
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: DYQAZJQDLPPHNB-UHFFFAOYSA-N
SMILES: CCCCC(=O)CC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)