missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-hexanone, 95%

Product Code. 11454363
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454363 25 g 25g
11434363 1 g 1g
11444363 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454363 Supplier Thermo Scientific Alfa Aesar Supplier No. A10944.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25870-62-6 | C12H16O | 176.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25870-62-6
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00027240
InChI Key DYQAZJQDLPPHNB-UHFFFAOYSA-N
Synonym 1-phenyl-2-hexanone, benzyl butyl ketone, 2-hexanone, 1-phenyl, benzyl n-butyl ketone, phenyl hexanone, benzylbutylketone, 1-phenyl-hexan-2-one, acmc-209gmx, dyqazjqdlpphnb-uhfffaoysa
PubChem CID 33191
IUPAC Name 1-phenylhexan-2-one
SMILES CCCCC(=O)CC1=CC=CC=C1

Specifications

Density 0.94
Boiling Point 153°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.501
Quantity 25 g
Beilstein 2044423
Formula Weight 176.26
Percent Purity 95%
Chemical Name or Material 1-Phenyl-2-hexanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.