accessibility menu, dialog, popup

UserName

H3CH3CO(R)O(S)O(R)OCH3CH3O(R)(R)HO

CAS RN 2595-05-3

IUPAC Name: (3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Synonyms: (3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
Molecular Weight (g/mol): 260.29
Molecular Formula: C12H20O6
InChi Key: KEJGAYKWRDILTF-VVULQXIFSA-N
SMILES: CC1(C)OC[C@@H](O1)[C@H]1O[C@@H]2OC(C)(C)O[C@@H]2[C@@H]1O
Molecular Weight (g/mol) 
  • (6)
  • (1)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results