missing translation for 'onlineSavingsMsg'
Learn More

1,2:5,6-Di-O-isopropylidene-alpha-D-allo-furanose, 98%, Thermo Scientific Chemicals

Product Code. 10521354 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10521354 10 g Glass bottle 10g
10219543 500 mg Glass bottle 500mg
10319440 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10521354 Supplier Thermo Scientific Acros Supplier No. 270960100
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2595-05-3 | Molecular Formula: C12H20O6 | Molecular Weight: 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2595-05-3
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.28
MDL Number MFCD00135634
InChI Key KEJGAYKWRDILTF-VVULQXIFSA-N
Synonym 1,2:5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyltetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2,5,6-di-o-isopropylidene-alpha-d-allofuranose, 3ar,5s,6r,6ar-5-4r-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol, 1,2:5,6-bis-o-1-methylethylidene, 5,6-di-o-isopropylidene-alpha-d-allofuranose, 1,2:5,6-di-o-isopropylidene-?-d-allofuranose, 1,2:5,6-di-o-isopropylidene-alpha-d-ribo-hexofuranose, 1-o,2-o:5-o,6-o-diisopropylidene-alpha-d-allofuranose
PubChem CID 7157054
IUPAC Name (3aR,5S,6R,6aR)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol
SMILES CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 73°C to 77°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Specific Rotation Condition +37.50 (20.00°C c=1,chloroform)
Specific Rotation + 37.5
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 2:5, 6-Di-O-isopropylidene-α-D-allo-furanose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.