CAS RN 2606-90-8
CAS RN 2606-90-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methyl-5-oxyethyl Thiazol Diphosphate Ammonium Salt, TRC
CAS: 2606-90-8 Molecular Formula: C6H11NO7P2S . xH3N Molecular Weight (g/mol): 303.17 Synonym: 2-(4-Methylthiazol-5-yl)ethyl Trihydrogen Diphosphate IUPAC Name: 2-(4-methyl-1,3-thiazol-5-yl)ethyl phosphono hydrogen phosphate SMILES: Cc1ncsc1CCOP(=O)(O)OP(=O)(O)O