missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-oxyethyl Thiazol Diphosphate Ammonium Salt, TRC

Product Code. 30474168
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474168 5 mg 5mg
30467644 25 mg 25mg
30466373 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474168 Supplier Toronto Research Chemicals Supplier No. TRCM6504105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2606-90-8
Molecular Formula C6H11NO7P2S . xH3N
Molecular Weight (g/mol) 303.17
Synonym 2-(4-Methylthiazol-5-yl)ethyl Trihydrogen Diphosphate
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethyl phosphono hydrogen phosphate
SMILES Cc1ncsc1CCOP(=O)(O)OP(=O)(O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H11NO7P2S/c1-5-6(17-4-7-5)2-3-13-16(11,12)14-15(8,9)10/h4H,2-3H2,1H3,(H,11,12)(H2,8,9,10)
Formula Weight 302.9731
Chemical Name or Material 4-Methyl-5-oxyethyl Thiazol Diphosphate Ammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.