accessibility menu, dialog, popup

UserName

CAS RN 26163-03-1

NH2NClBr

CAS RN 26163-03-1

IUPAC Name: 3-bromo-5-chloropyridin-2-amine
Synonyms: 3-bromo-5-chloropyridin-2-amine
Molecular Weight (g/mol): 207.46
Molecular Formula: C5H4BrClN2
InChi Key: UWGGGYYCKDCTGN-UHFFFAOYSA-N
SMILES: NC1=NC=C(Cl)C=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)