missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-chloropyridine, 95%, Thermo Scientific™

Product Code. 15433787
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433787 10 g 10g
11372999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433787 Supplier Thermo Scientific Alfa Aesar Supplier No. H27799.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 26163-03-1
Molecular Formula C5H4BrClN2
Molecular Weight (g/mol) 207.455
MDL Number MFCD00955627
InChI Key UWGGGYYCKDCTGN-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-chloropyridine, 3-bromo-5-chloro-2-pyridinamine, 2-pyridinamine, 3-bromo-5-chloro, 3-bromo-5-chloro-2-pyridylamine, 3-bromo-5-chloropyrid-2-ylamine, 3-bromo-5-chloro-pyridin-2-ylamine, abbypharma ap-10-5406, 3-bromo-5-chloro-pyridin-2-amine, pubchem1141, acmc-1ch3b
PubChem CID 2763971
IUPAC Name 3-bromo-5-chloropyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)Cl

Spezifikation

Melting Point 83°C to 85°C
Quantity 10 g
Formula Weight 207.46
Percent Purity 95%
Chemical Name or Material 2-Amino-3-bromo-5-chloropyridine
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.