accessibility menu, dialog, popup

UserName

CAS RN 261762-35-0

IUPAC Name: 5-bromo-1,2-difluoro-3-methoxybenzene
Synonyms: 5-bromo-1,2-difluoro-3-methoxybenzene
Molecular Weight (g/mol): 223.02
Molecular Formula: C7H5BrF2O
InChi Key: UDTVFZBQFYOZST-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results