accessibility menu, dialog, popup

UserName

CAS RN 261762-41-8

IUPAC Name: 3-chloro-2,6-difluorobenzamide
Synonyms: 3-chloro-2,6-difluorobenzamide
Molecular Weight (g/mol): 191.56
Molecular Formula: C7H4ClF2NO
InChi Key: TUTOELHQVJTZTO-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C=CC(Cl)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results