accessibility menu, dialog, popup

UserName

CAS RN 261762-88-3

IUPAC Name: 2-(bromomethyl)-1-chloro-3-fluoro-4-methylbenzene
Synonyms: 2-(bromomethyl)-1-chloro-3-fluoro-4-methylbenzene
Molecular Weight (g/mol): 237.5
Molecular Formula: C8H7BrClF
InChi Key: QWQVTYYKMSFGGP-UHFFFAOYSA-N
SMILES: CC1=C(F)C(CBr)=C(Cl)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results