accessibility menu, dialog, popup

UserName

CAS RN 261762-91-8

CH3ClHOF

CAS RN 261762-91-8

IUPAC Name: 6-chloro-2-fluoro-3-methylphenol
Synonyms: 6-chloro-2-fluoro-3-methylphenol
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: HCZRPIVKIPIGDM-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C(O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results