accessibility menu, dialog, popup

UserName

FFFBrCl

CAS RN 261763-22-8

IUPAC Name: 1-(bromomethyl)-2-chloro-3-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-2-chloro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 273.48
Molecular Formula: C8H5BrClF3
InChi Key: ZPHUUSQDKIEPDN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(CBr)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results