accessibility menu, dialog, popup

UserName

CH3NH2FF

CAS RN 261763-41-1

IUPAC Name: 1-(2,3-difluoro-4-methylphenyl)methanamine
Synonyms: 1-(2,3-difluoro-4-methylphenyl)methanamine
Molecular Weight (g/mol): 157.16
Molecular Formula: C8H9F2N
InChi Key: BHMZMUKPFBBPKG-UHFFFAOYSA-N
SMILES: CC1=C(F)C(F)=C(CN)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results