accessibility menu, dialog, popup

UserName

CAS RN 261951-71-7

CH3FBr

CAS RN 261951-71-7

IUPAC Name: 2-(bromomethyl)-4-fluoro-1-methylbenzene
Synonyms: 2-(bromomethyl)-4-fluoro-1-methylbenzene
Molecular Weight (g/mol): 203.05
Molecular Formula: C8H8BrF
InChi Key: DRIYGEORZARQGO-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1CBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results