accessibility menu, dialog, popup

UserName

CH3H3CCH3OON(S)OHO

CAS RN 26250-84-0

IUPAC Name: (2S)-1-[(tert-butoxy)carbonyl]piperidine-2-carboxylic acid
Synonyms: (2S)-1-(tert-butoxycarbonyl)piperidine-2-carboxylic acid
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: JQAOHGMPAAWWQO-QMMMGPOBSA-N
SMILES: CC(C)(C)OC(=O)N1CCCC[C@H]1C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (5)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)