missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-pipecolinic acid, 98+%

Product Code. 11384286
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384286 5 g 5g
11394286 250 mg 250mg
11374286 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384286 Supplier Thermo Scientific Alfa Aesar Supplier No. L19361.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26250-84-0 | C11H19NO4 | 229.276 g/mol

N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26250-84-0
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00151904
InChI Key JQAOHGMPAAWWQO-QMMMGPOBSA-N
Synonym boc-pip-oh, s-1-boc-piperidine-2-carboxylic acid, s-n-boc-piperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-l-pipecolic acid, boc-l-pip-oh, n-boc-l-pipecolinic acid, l-n-boc-pipecolic acid, 2s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl piperidine-2-carboxylic acid
PubChem CID 688617
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O

Specifications

Melting Point 121°C to 125°C
Quantity 5 g
Beilstein 3652038
Solubility Information Insoluble in water.
Optical Rotation −46° (c=1 in Methanol)
Formula Weight 229.28
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-pipecolinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.