accessibility menu, dialog, popup

UserName

OONH2(S)H2NHClHClCH3

CAS RN 26348-70-9

IUPAC Name: dihydrogen methyl (2S)-2,6-diaminohexanoate dichloride
Synonyms: dihydrogen L-lysine methyl ester dichloride
Molecular Weight (g/mol): 233.13
Molecular Formula: C7H18Cl2N2O2
InChi Key: SXZCBVCQHOJXDR-ILKKLZGPSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)