missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Methyl Ester Dihydrochloride, TRC

Product Code. 30483833
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483833 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483833 Supplier Toronto Research Chemicals Supplier No. TRCL4689005G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Lysine Methyl Ester Dihydrochloride
CAS 26348-70-9
Molecular Formula C7H18Cl2N2O2
Quantity 5 g
Synonym L-Lysine Methyl Ester DiHydrochloride, Methyl L-Lysinate Hydrochloride, NSC 83256
InChI Formula InChI=1S/C7H16N2O2.2ClH/c1-11-7(10)6(9)4-2-3-5-8;;/h6H,2-5,8-9H2,1H3;2*1H/t6-;;/m0../s1
SMILES Cl.Cl.COC(=O)[C@@H](N)CCCCN
IUPAC Name methyl (2S)-2,6-diaminohexanoate;dihydrochloride
Molecular Weight (g/mol) 233.14
Formula Weight 232.0745
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.