accessibility menu, dialog, popup

UserName

SSOOOOONOHNHCH3H3CCH3H3CNa(Z)(Z)

CAS RN 2650-17-1

IUPAC Name: sodium 4-{[(1E)-4-({2-[4-(ethylamino)-3-methylphenyl]ethyl}imino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
Synonyms: xylene cyanol
Molecular Weight (g/mol): 538.61
Molecular Formula: C25H27N2NaO6S2
InChi Key: UQGVMAOBDMCVOQ-PULQMKEHSA-M
SMILES: [Na+].CCNC1=CC=C(CCN=C2C=C\C(=C/C3=CC=C(C=C3S(O)(=O)=O)S([O-])(=O)=O)C=C2C)C=C1C
Quantity 
  • (6)
  • (3)
Color 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (6)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)