missing translation for 'onlineSavingsMsg'
Learn More

Xylene Cyanole FF (Bluish-Green Powder/Electrophoresis), Fisher BioReagents

Product Code. 10532965
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10532965 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10532965 Supplier Fisher BioReagents Supplier No. BP56510

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This dye is used in electrophoretic applications to mark the migration position of nucleic acid fragments.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 295°C
CAS 2650-17-1
Molecular Formula C25H27N2NaO6S2
MDL Number MFCD00019481
InChI Key VVLFAAMTGMGYBS-KRQUPCAFSA-M
SMILES [Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O
IUPAC Name sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
Molecular Weight (g/mol) 538.61
Lambda Max 612 to 616nm
Formula Weight 538.61
Loss on Drying 10% max.
Grade Electrophoresis
Molar Absorption Coefficient 30000 min.
Packaging Glass Bottle
Color Index No. 42135
Color Green
pH 3.8
Quantity 10 g
Physical Form Solid
Chemical Name or Material Xylene Cyanole FF
Show More Show Less
compliance-icons
Product Identifier
  • Xylene cyanol FF
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.