accessibility menu, dialog, popup

UserName

CAS RN 267006-24-6

BFFFFK

CAS RN 267006-24-6

IUPAC Name: potassium trifluoro(3-fluorophenyl)boranuide
Synonyms: potassium trifluoro(3-fluorophenyl)boranuide
Molecular Weight (g/mol): 202
Molecular Formula: C6H4BF4K
InChi Key: LOPFKHUYBDOHRD-UHFFFAOYSA-N
SMILES: [K+].FC1=CC=CC(=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results