accessibility menu, dialog, popup

UserName

CAS RN 267006-26-8

BFFFFFK

CAS RN 267006-26-8

IUPAC Name: potassium (3,5-difluorophenyl)trifluoroboranuide
Synonyms: potassium (3,5-difluorophenyl)trifluoroboranuide
Molecular Weight (g/mol): 219.99
Molecular Formula: C6H3BF5K
InChi Key: JFSPHPHYZHGKPO-UHFFFAOYSA-N
SMILES: [K+].FC1=CC(=CC(F)=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results