accessibility menu, dialog, popup

UserName

H3COBrOCH3Br

CAS RN 2674-34-2

IUPAC Name: 1,4-dibromo-2,5-dimethoxybenzene
Synonyms: 1,4-dibromo-2,5-dimethoxybenzene
Molecular Weight (g/mol): 295.96
Molecular Formula: C8H8Br2O2
InChi Key: CHCLRVOURKGRSW-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(OC)C=C1Br
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results