accessibility menu, dialog, popup

UserName

OON

CAS RN 27018-76-4

IUPAC Name: 1-benzyl-1H-indole-3-carboxylate
Synonyms: 1-benzylindole-3-carboxylate
Molecular Weight (g/mol): 250.28
Molecular Formula: C16H12NO2
InChi Key: LVYDDRHDOKXFMW-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CN(CC2=CC=CC=C2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results