missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylindole-3-carboxylic acid, 98%, Thermo Scientific™

Product Code. 10527990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10527990 5 g Glass bottle 5g
10008249 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10527990 Supplier Thermo Scientific Acros Supplier No. 428260050
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 27018-76-4
Molecular Formula C16H13NO2
Molecular Weight (g/mol) 251.28
MDL Number MFCD00057094
InChI Key LVYDDRHDOKXFMW-UHFFFAOYSA-N
Synonym 1-benzyl-1h-indole-3-carboxylic acid, indole-3-carboxylic acid, 1-benzyl, 1h-indole-3-carboxylic acid, 1-phenylmethyl, pubchem7313, cbmicro_033327, cambridge id 5791025, 5-22-03-00037 beilstein handbook reference, 1-benzylindol-3-carboxylic acid, n-benzylindole-3-carboxylic acid, 1h-indole-3-carboxylicacid, 1-phenylmethyl
PubChem CID 33671
IUPAC Name 1-benzylindole-3-carboxylic acid
SMILES C1=CC=C(C=C1)CN2C=C(C3=CC=CC=C32)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 251.28
Percent Purity 98%
Chemical Name or Material 1-Benzylindole-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.