accessibility menu, dialog, popup

UserName

CH3CH3Br

CAS RN 27129-86-8

IUPAC Name: 1-(bromomethyl)-3,5-dimethylbenzene
Synonyms: 1-(bromomethyl)-3,5-dimethylbenzene
Molecular Weight (g/mol): 199.09
Molecular Formula: C9H11Br
InChi Key: QXDHXCVJGBTQMK-UHFFFAOYSA-N
SMILES: CC1=CC(CBr)=CC(C)=C1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)