CAS RN 2714485-34-2
CAS RN 2714485-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac Phenylephrine-d3 Hydrochloride, TRC
CAS: 2714485-34-2 Molecular Formula: C9H10D3NO2 . HCl Molecular Weight (g/mol): 206.69 Synonym: (±)-3-Hydroxy-α-[(methylamino)methyl]benzenemethanol-d3 Hydrochloride,,(±)-m-Hydroxy-α-[(methylamino)methyl]benzyl Alcohol-d3 Hydrochloride,(±)-Phenylephrine Hydrochloride-d3,1-(3-Hydroxyphenyl)-2-methylaminoethanol-d3 Hydrochloride,DL-Phenylephrine-d3 Hydrochloride,NSC 14029-d3 SMILES: OC1=CC=CC(C(O)CNC([2H])([2H])[2H])=C1.Cl