missing translation for 'onlineSavingsMsg'
Learn More

rac Phenylephrine-d3 Hydrochloride, TRC

Product Code. 30417007
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417007 2.5 mg 2.5mg
30408915 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30417007 Supplier Toronto Research Chemicals Supplier No. TRCP3206372.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2714485-34-2
Molecular Formula C9H10D3NO2 . HCl
Molecular Weight (g/mol) 206.69
Synonym (±)-3-Hydroxy-α-[(methylamino)methyl]benzenemethanol-d3 Hydrochloride,, (±)-m-Hydroxy-α-[(methylamino)methyl]benzyl Alcohol-d3 Hydrochloride, (±)-Phenylephrine Hydrochloride-d3, 1-(3-Hydroxyphenyl)-2-methylaminoethanol-d3 Hydrochloride, DL-Phenylephrine-d3 Hydrochloride, NSC 14029-d3
SMILES OC1=CC=CC(C(O)CNC([2H])([2H])[2H])=C1.Cl

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C9H13NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,9-12H,6H2,1H3;1H/i1D3;
Formula Weight 206.09
Chemical Name or Material rac Phenylephrine-d3 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.