accessibility menu, dialog, popup

UserName

CAS RN 2725-81-7

ONOOO

CAS RN 2725-81-7

IUPAC Name: 6-nitro-2H-chromen-2-one
Synonyms: 6-nitrochromen-2-one
Molecular Weight (g/mol): 191.14
Molecular Formula: C9H5NO4
InChi Key: RMERXEXZXIVNBF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2OC(=O)C=CC2=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)