accessibility menu, dialog, popup

UserName

CAS RN 2732-80-1

H3COClBr

CAS RN 2732-80-1

IUPAC Name: 2-bromo-1-chloro-4-methoxybenzene
Synonyms: 2-bromo-1-chloro-4-methoxybenzene
Molecular Weight (g/mol): 221.48
Molecular Formula: C7H6BrClO
InChi Key: SQHMXVXKKCXIGN-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results